missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Cortistatin 14
GREENER_CHOICE

Product Code. 18023802
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18023802

Brand: Tocris Bioscience™ 3374/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Endogenous neuropeptide; binds sst1 - sst5, ghrelin receptor and MRGPRX2

Cortistatin 14 is an endogenous neuropeptide, mainly expressed in the cortex and hippocampus, that has structural and functional similarities to somatostatin-14. Cortistatin 14 displays potency at all somatostatin receptors (sst1 - sst5) and prevents somatostatin-14 binding (IC50 values are 0.09, 0.2, 0.3, 0.3 and 5 nM at sst2, sst4, sst3, sst5 and sst1 receptors, respectively). Also a potent MRGPRX2 agonist (EC50 = 25 nM in HEK293 cells) and binds to the growth hormone secretagog receptor (GHS-R1a). Exhibits neuronal depressant and sleep-modulating properties in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Purity 0.95
Content And Storage Store at -20°C
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.