Learn More
Tocris Bioscience™ Amylin
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Description
Amylin is an endogenous peptide agonist for amylin, calcitonin, CGRP and adrenomedullin receptors. Inhibits glucagon secretion, delays gastric emptying and acts as a satiety agent. Displays glucose lowering effects in vivo.
Specifications
Specifications
| Quantity | 500 μg |