Learn More
Tocris Bioscience™ ACTH (1-39)
GREENER_CHOICE
Product Code. 18055482
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 18055482
Brand: Tocris Bioscience™ 3492/1
Potent endogenous MC2 agonist
ACTH (1-39) is a potent endogenous melanocortin receptor 2 (MC2) agonist (EC50 = 57 pM). Component of the hypothalamic-pituitary-adrenal (HPA) axis that stimulates glucocorticoid production and release from the adrenal cortex. Induces insulin resistance, promotes a proinflammatory profile and stimulates UCP-1 in adipocytes in vitro.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg