missing translation for 'onlineSavingsMsg'
Learn More

Tocofersolan, MedChemExpress

Product Code. p-200096768
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
This item is not returnable. View return policy

Product Code. 16438588

Brand: Medchem Express HYB0717

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tocofersolan is synthetic polyethylene glycol derivative of α-tocopherol. Tocofersolan is an orally active and water-soluble analog of vitamin E. Tocofersolan can reduce neurobehavioral deficits in zebrafish embryos exposed to moderate and high concentrations of BaP during early development. Tocofersolan shows antioxidant activity. Tocofersolan can be used to provide an orally bioavailable source of vitamin E.

Tocofersolan is synthetic polyethylene glycol derivative of α-tocopherol. Tocofersolan is an orally active and water-soluble analog of vitamin E. Tocofersolan can reduce neurobehavioral deficits in zebrafish embryos exposed to moderate and high concentrations of BaP during early development. Tocofersolan shows antioxidant activity. Tocofersolan can be used to provide an orally bioavailable source of vitamin E.

Specifications

Chemical Name or Material Tocofersolan
CAS 9002-96-4
Color Off-White
Purity Grade Notes Research
Quantity 10 g
Synonym TPGS D-α-Tocopherol polyethylene glycol 1000 succinate Vitamin E-TPGS
Solubility Information H2O : 100 mg/mL (66.09 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (66.09 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (33.05 mM; Need ultrasonic)
SMILES CC1=C(CCC(CCCC(C)CCCC(C)CCCC(C)C)(C)O2)C2=C(C)C(C)=C1OC(CCC(OCCO)=O)=O.[n]
Molecular Weight (g/mol) 1513.0
Formula Weight 1513.0
Percent Purity 96.0%
Grade Research
Physical Form Oil
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.