missing translation for 'onlineSavingsMsg'
Learn More

Fisher Chemical™ TOC Standard Solution 10 mg/L

Product Code. 31034050 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31034050 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 31034050 Supplier Fisher Chemical™ Supplier No. CM 28225
  • ONLINE EXCLUSIVE PRICE
    £137.00
    £117.14 / 100mL
    Save £19.86
    14% Off
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Known-carbon reference solutions used to calibrate TOC analyzers, ensuring accurate measurement of organic carbon in water samples.

  • Accuracy: Provide a reliable baseline for measuring TOC in environmental, pharmaceutical, and industrial water samples.
  • Quality control: Required in regulated industries (e.g., USP for pharmaceutical water).
  • Traceability: This TOC standards is certified ISO 17025 and ISO 17034 and traceable to NIST (National Institute of Standards and Technology).
  • Each CRM comes with a certificate of analysis that includes uncertainty calculations.
  • This documentation is critical for audits, method validation, and proving the reliability of your data.

Specifications

Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.