missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TOC Certified Containers

Trust our low-level certified vials for Total Organic Carbon testing and sampling. TOC-certified products are manufactured, packaged and tested to exacting standards.

Brand:  Thermo Scientific™ 3115-OTWS-2

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 15153488

  • £1966.21 / Pack of 100
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

Trust our low-level certified vials for Total Organic Carbon testing and sampling. Manufactured, tested and packaged by exacting standards,Thermo Scientific™ TOC Certified Containers are certified to contribute <10ppb or < 20ppb TOC as background.

Containers:
  • Available in several sizes and styles of containers, including the popular 40mL autosampler vials cleaned, certified and ready to use
  • Include Certificate of Analysis
Includes:
  • Sampling and storage containers
  • Autosampler compatible vials
  • Culture tubes
  • Low TOC water
Recommended for:
  • TOC testing
  • USP method 643 testing
  • Off-line grab sampling of high purity water
  • Storage of validation standards.

Specifications

Specifications

Polysulphone Tube, TOC Cleaned
1.02 in.
Polysulfone
3.74 in.
1 oz.
95 mm
Certified to contribute <10ppb, <20ppb, Certificate of Analysis (COA), Certificate of Conformance (COC)
20-400
PTFE/Silicone
26 mm
30 mL
100
30 mL
100 Cs.
Container
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers