missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TOC Certified Containers

Trust our low-level certified vials for Total Organic Carbon testing and sampling. TOC-certified products are manufactured, packaged and tested to exacting standards.

Brand:  Thermo Scientific™ 1000A/TOC

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 15133488

  • £185.00 / Pack of 12
Estimated Shipment: 12-09-2024
to see stock.
Add to basket

Description

Description

Trust our low-level certified vials for Total Organic Carbon testing and sampling. Manufactured, tested and packaged by exacting standards,Thermo Scientific™ TOC Certified Containers are certified to contribute <10ppb or < 20ppb TOC as background.

Containers:
  • Available in several sizes and styles of containers, including the popular 40mL autosampler vials cleaned, certified and ready to use
  • Include Certificate of Analysis
Includes:
  • Sampling and storage containers
  • Autosampler compatible vials
  • Culture tubes
  • Low TOC water
Recommended for:
  • TOC testing
  • USP method 643 testing
  • Off-line grab sampling of high purity water
  • Storage of validation standards.

Specifications

Specifications

Amber Boston round glass bottle with closure
3.75 in.
95.25 mm
1000 mL
12
1000 mL
12 Cs.
Container
PTFE
Amber
Glass
8.5 in.
32 oz.
215.9 mm
Certified to contribute <10ppb, <20ppb, Certificate of Analysis (COA), Certificate of Conformance (COC)
33-430
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers