missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin sulfate, Thermo Scientific™

A water-soluble aminoglycoside antibiotic

Brand:  Thermo Scientific Alfa Aesar J62995.03

Quantity 1g

 View more versions of this product

Product Code. 15405279

  • £273.00 / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Tobramycin sulfate is used as an antibiotic which is used to treat various types of bacterial infections, particularly Gram-negative infections. It is especially effective against species of Pseudomonas. It is used to inhibit bacterial protein synthesis at the level of 30S (16S rRNA) and 70S ribosomal complex assembly. It is used to treat pseudomonas aeruginosa lung infections and is used in combination with other antibiotics to treat urinary tract infections, gynecologic infections, peritonitis, endocarditis, pneumonia, sepsis, respiratory infections, osteomyelitis and other soft-tissue infections. It is a potential therapy for sinus infections1. Product T1783 has been used to study antibiotic resistance2.

Solubility
Soluble in water (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

49842-07-1
565.59
ZEUUPKVZFKBXPW-UHFFFAOYNA-N
4-amino-2-[(4,6-diamino-3-{[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-6-(hydroxymethyl)oxane-3,5-diol; sulfuric acid
C18H39N5O13S
MFCD00133864,MFCD00133864
54413
OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(N)CC1O

Specifications

49842-07-1
1g
MFCD00133864,MFCD00133864
Soluble in water (50mg/ml).
OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(N)CC1O
565.59
1425.41
White
C18H39N5O13S
14,9490
ZEUUPKVZFKBXPW-UHFFFAOYNA-N
4-amino-2-[(4,6-diamino-3-{[3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]-6-(hydroxymethyl)oxane-3,5-diol; sulfuric acid
54413
Tobramycin sulfate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.