missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tobramycin Deuterated, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Tobramycin Deuterated |
| Molecular Formula | C18H37N5O9 for undeuterated |
| Quantity | 10 mg |
| Synonym | O-3-Amino-3-deoxy-α-D-glucopyranosyl-(1-6)-O-[2,6-diamino-2,3,6-trideoxy-α -D-ribo-hexopyranosyl-(1-4)]-2-deoxy-D-streptamine Deuterated, Nebramycin 6- Deuterated, Nebramycin VI- Deuterated, Tobramax-Deuterated, Tobramaxin- Deuterated, Tobrex-Deuterated |
| InChI Formula | InChI=1S/C18H37N5O9/c19-3-9-8(25)2-7(22)17(29-9)31-15-5(20)1-6(21)16(14(15)28)32-18-13(27)11(23)12(26)10(4-24)30-18/h5-18,24-28H,1-4,19-23H2/t5-,6+,7+,8-,9+,10+,11-,12+,13+,14-,15+,16-,17+,18+/m0/s1 |
| SMILES | N[C@@H]1[C@@H](O[C@H]2O[C@H](CN)[C@@H](O)C[C@H]2N)[C@H](O)[C@@H](O[C@H]3O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H](N)C1 |
| Molecular Weight (g/mol) | 467.51 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction