missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin Carbamate Acetate, Tobramycin Carbamate Acetate, TRC

Product Code. 30008691
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008691 10 mg 10mg
30005135 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008691 Supplier Toronto Research Chemicals Supplier No. TRCT52402010MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C19H38N6O10 xC2H4O2
Molecular Weight (g/mol) 510.54
Synonym O-3-Amino-6-O-(aminocarbonyl)-3-deoxy-α-D-glucopyranosyl-(1→6)-O-[2,6-diamino-2,3,6-trideoxy-α-D-ribo-hexopyranosyl-(1→4)]-2-deoxy-D-streptamine, 6' '-O-Carbamoyltobramycin Acetate, Nebramycin V' Acetate, Nebramycin factor 5' Acetate
SMILES OC(C)=O.N[C@@H]1C[C@H](O)[C@@H](CN)O[C@@H]1O[C@H]2[C@H](O)[C@@H](O[C@H]3O[C@H](COC(N)=O)[C@@H](O)[C@H](N)[C@H]3O)[C@H](N)C[C@@H]2N

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H38N6O10.C2H4O2/c20-3-9-8(26)2-7(23)17(32-9)34-15-5(21)1-6(22)16(14(15)29)35-18-13(28)11(24)12(27)10(33-18)4-31-19(25)30;1-2(3)4/h5-18,26-29H,1-4,20-24H2,(H2,25,30);1H3,(H,3,4)/t5-,6+,7+,8-,9+,10+,11-,12+,13+,14-,15+,16-,17+,18+;/m0./s1
Chemical Name or Material Tobramycin Carbamate Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.