missing translation for 'onlineSavingsMsg'
Learn More

Tobramycin, 900μg/mg

Product Code. 15457239
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457239 5 g 5g
15447239 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457239 Supplier Thermo Scientific Alfa Aesar Supplier No. J66040.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32986-56-4 | C18H37N5O9 | 467.52 g/mol

Primarily targets Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32986-56-4
Molecular Formula C18H37N5O9
Molecular Weight (g/mol) 467.52
MDL Number MFCD00077885
InChI Key NLVFBUXFDBBNBW-PBSUHMDJSA-N
Synonym tobramycin, nebramycin 6, nebramycin factor 6, nebramycin vi, tobramicin, tobramycetin, nebcin, 3'-deoxykanamycin b, aktob, tobi
PubChem CID 36294
ChEBI CHEBI:28864
IUPAC Name (2S,3R,4S,5S,6R)-4-amino-2-[(1S,2S,3R,4S,6R)-4,6-diamino-3-[(2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxyoxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-6-(hydroxymethyl)oxane-3,5-diol
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(CC(C(O3)CN)O)N)N

Specifications

Packaging Plastic bottle
Quantity 5 g
Beilstein 1357507
Solubility Information Soluble in water
Formula Weight 467.51
Physical Form Powder
Chemical Name or Material Tobramycin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.