missing translation for 'onlineSavingsMsg'
Learn More

Melford Tobramycin

Product Code. 30018871
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018871 5 g 5g
30018870 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30018871 Supplier Melford Supplier No. T521505.0
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tobramycin is an antibiotic that belongs to the aminoglycoside class of antibiotics and is primarily employed to treat various bacterial infections.

Tobramycin is an antibiotic that belongs to the aminoglycoside class of antibiotics and is primarily employed to treat various bacterial infections. It is effective against a range of gram-negative bacteria.

Tobramycin works by inhibiting bacterial protein synthesis. It binds to the bacterial ribosomes, disrupting the synthesis of proteins essential for bacterial growth and survival.

It is particularly effective against Gram-negative bacteria, including Pseudomonas aeruginosa, Escherichia coli, Klebsiella pneumoniae, and others. It is less effective against Gram-positive bacteria.

Specifications

Chemical Name or Material Tobramycin
CAS 32986-56-4
Quantity 5 g
Molecular Formula C18H37N5O9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.