missing translation for 'onlineSavingsMsg'
Learn More

Melford Tobramycin

Product Code. 30018870
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 30018870

Brand: Melford T521501.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tobramycin is an antibiotic that belongs to the aminoglycoside class of antibiotics and is primarily employed to treat various bacterial infections.

Tobramycin is an antibiotic that belongs to the aminoglycoside class of antibiotics and is primarily employed to treat various bacterial infections. It is effective against a range of gram-negative bacteria.

Tobramycin works by inhibiting bacterial protein synthesis. It binds to the bacterial ribosomes, disrupting the synthesis of proteins essential for bacterial growth and survival.

It is particularly effective against Gram-negative bacteria, including Pseudomonas aeruginosa, Escherichia coli, Klebsiella pneumoniae, and others. It is less effective against Gram-positive bacteria.

Specifications

Chemical Name or Material Tobramycin
CAS 32986-56-4
Quantity 1 g
Molecular Formula C18H37N5O9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.