Learn More
Novus Biologicals™ TOB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30840-100ug
 View more versions of this product
Product Code. 30067230
  • £386.00 / 100µg
Description
TOB1 Polyclonal antibody specifically detects TOB1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TOB1
Polyclonal
Western Blot : 1-5 μg/mL
APRO6, MGC104792, PIG49, protein Tob1, TOBMGC34446, Transducer of erbB-2 1, transducer of ERBB2, 1, TROB, TROB1
Synthetic peptide derived from the TOB1
100 μg
Cell Cycle and Replication
10140
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Human
Purified