missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNXB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580173
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: MCF-7 whole cell, SW620 whole cell, HELA whole cell. IHC: human placenta tissue. ICC/IF: MCF7 cell. Flow: Hela cell.
Tenascin-X (TNXB) is a extracellular matrix glycoprotein predominantly located in the outer reticular lamina of the basement membrane (BM). It interacts with many other ECM proteins and it accelerates collagen fibrillogenesisin vitro. TNXB plays a role in interactions between cell and ECM, antiadhesive effect, inhibiting cell migration, and in maintaining homeostasis of the extracellular matrix. Deficiency of TNXB has been associated with the connective tissue disorder Ehlers-Danlos syndrome.
Specifications
| TNXB | |
| Polyclonal | |
| Unconjugated | |
| TNXB | |
| EDS3; growth-inhibiting protein 45; hexabrachion-like protein; HXBL; tenascin XB; tenascin XB1; tenascin XB2; tenascin-X; TENX; Tn-mhc; Tnx; TN-X; TNXB; TNXB1; TNXB2; TNXB2tenascin XB1; TNXBS; TNXBSTNXTENX; VUR8; XB; XBS; XB-S; XBTN-X | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7148 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| P22105 | |
| TNXB | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human TNXB (3309-3337aa DLRAVAVSGLDPARKYKFLLFGLQNGKRH). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction