missing translation for 'onlineSavingsMsg'
Learn More

TNS1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16819544 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16819544 20 μL 20µL
16809544 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16819544 Supplier Proteintech Supplier No. 200541AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody recognizes a region near the N-terminus of TNS1.

Tensin is involved in the maintece of cellular structure by anchoring Actin filaments at the focal adhesion via F-Actin binding and capping activities. However, tensin also contains a Src homology 2 (SH2) domain and has the ability to be phosphorylated. Tensin is phosphorylated on tyrosine, serine, and threonine residues, suggesting that it might participate in signal transduction cascades. These diverse characteristics in a single molecule indicate that tensin may be an important link between the cytoskeleton and signal transduction pathways.

Specifications

Antigen TNS1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TNS1
Gene Accession No. Q9HBL0
Gene Alias MST091, MST122, MST127, MSTP122, MSTP127, MXRA6, tensin 1, TNS, TNS1
Gene Symbols TNS1
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21961, 7145
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.