missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNPO3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 12818732
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12818732 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12818732 Supplier Invitrogen™ Supplier No. PA520551

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat liver tissue lysate. PA5-20551 can be used with blocking peptide PEP-0671.

TNPO3 is a member of the importin beta/transportin family of nuclear import receptors and is specific for the essential splicing factors termed SR proteins, binding specifically and directly to the arginine/serine (RS)-rich domains of these proteins. Further experiments demonstrated that this interaction occurs only when these RS domains are phosphorylated. More recently, TNPO3 was identified as an HIV dependency factor (HDF) and that depletion of TNPO3 resulted in HIV inhibition after reverse transcription but before integration, suggesting that TNPO3 may be an important drug target in HIV treatment. At least three isoforms of TNPO3 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNPO3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TNPO3
Gene Accession No. Q6P2B1, Q9Y5L0
Gene Alias 5730544L10Rik; C430013M08Rik; C81142; D6Ertd313e; Imp12; importin 12; Importin-12; IPO12; LGMD1F; limb girdle muscular dystrophy 1F (autosomal dominant); mKIAA4133; MTR10A; Tnpo3; transportin 3; transportin-3; Transportin-SR; TRNSR; TRN-SR; TRN-SR2
Gene Symbols TNPO3
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the amino terminus of human TNPO3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23534, 296954, 320938
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.