missing translation for 'onlineSavingsMsg'
Learn More

TNP2 Polyclonal Antibody
GREENER_CHOICE

TNP2 Polyclonal Antibody

Brand:  Invitrogen PA577020

Product Code. 15933535

  • £472.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TNP2 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TNP2
Polyclonal
Unconjugated
TNP2
haploid expressed, male germcell specific, chromatin binding; nuclear transition protein 2; TNP2; TP2; TP-2; transition protein 2; transition protein 2 (during histone to protamine replacement)
Rabbit
Antigen Affinity Chromatography
RUO
7142
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Q05952
TNP2
Recombinant full length Human TNP2.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.