missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17178-1-AP-150UL
This item is not returnable.
View return policy
Description
TNP1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines. Transition protein-1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines (see PRM1, MIM 182880; PRM2, MIM 182890) (Luerssen et al., 1990 ). Sequence Note: removed 4 bases from the 5′ end that did not align to the reference genome assembly.Specifications
| TNP1 | |
| Polyclonal | |
| Unconjugated | |
| TNP1 | |
| STP 1, TNP1, TP 1, TP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21958, 24839, 7141 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02317, P09430, P10856 | |
| TNP1 | |
| TNP1 Fusion Protein Ag10890 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction