missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100119
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TNP1.
TNP1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines. Transition protein-1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines (see PRM1, MIM 182880; PRM2, MIM 182890) (Luerssen et al., 1990 ). Sequence Note: removed 4 bases from the 5' end that did not align to the reference genome assembly.
Specifications
| TNP1 | |
| Polyclonal | |
| Unconjugated | |
| Tnp1 | |
| Protein T; Spermatid nuclear transition protein 1; STP-1; Testis-specific basic protein; tnp; TNP1; TNP1 protein; TP1; TP-1; transition protein 1; transition protein 1 (during histone to protamine replacement) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21958, 24839, 7141 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P02317, P09430, P10856 | |
| Tnp1 | |
| A synthesized peptide derived from human TNP1(Accession P09430), corresponding to amino acid residues T3-S53. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction