missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNP1, Mouse anti-Human, Clone: 1B5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00007141-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MSTSRKLKSHGMRRSKSRSPHKGVKRGGSKRKYRKGNLKSRKRGDDANRNYRSHLSpecifications
| TNP1 | |
| Monoclonal | |
| Unconjugated | |
| transition protein 1 (during histone to protamine replacement) | |
| TP1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1B5 | |
| In 1x PBS, pH 7.4 | |
| BC029516 | |
| TNP1 | |
| TNP1 (AAH29516.1, 1 a.a. ∼ 55 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 7141 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction