missing translation for 'onlineSavingsMsg'
Learn More

TNP-470, MedChemExpress

Product Code. 16476377
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476377 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16476377 Supplier Medchem Express Supplier No. HY101932
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TNP-470 is a methionine aminopeptidase-2 inhibitor and also an angiogenesis inhibitor.

TNP-470 is a methionine aminopeptidase-2 inhibitor and also an angiogenesis inhibitor.

Specifications

Chemical Name or Material TNP-470
CAS 129298-91-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H28ClNO6
Quantity 5 mg
Synonym AGM-1470
Solubility Information DMSO : 100 mg/mL (248.83 mM; Need ultrasonic)
SMILES C[C@]1([C@@H](C/C=C(C)/C)O1)[C@]([C@@H]2OC)([H])[C@]3(CC[C@H]2OC(NC(CCl)=O)=O)CO3
Molecular Weight (g/mol) 401.88
Formula Weight 401.88
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.