missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNNT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563365
This item is not returnable.
View return policy
Description
Immunogen sequence: EEEAQEEAAE VHEEVHEPEE VQEDTAEEDA EEEKPRPKLT APK Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.
Specifications
| TNNT3 | |
| Polyclonal | |
| Unconjugated | |
| TNNT3 | |
| beta TnTF; beta-TnTF; Fast skeletal muscle troponin T; fast skeletal muscle troponin T3; fast skeletal troponin T beta-1 isoform; fetal troponin T 1; fetal troponin T 2; fetal troponin T 3; fTnT; skeletal muscle fast-twitch TnT; skeletal troponin T alpha-1 isoform; Tnnt3; Tnt; TnT1f; TnT2.5f; TnT2f; TnT2fa; TnT4f; tnTf; troponin T 2.5f precursor; troponin T 2f precursor; troponin T 2fa precursor; troponin T 3f; troponin T 4f precursor; troponin T fast skeletal muscle type; troponin T type 3 (skeletal, fast); troponin T, fast skeletal muscle; troponin T1f precursor; troponin T3, fast skeletal type; troponin T3, skeletal, fast; troponin-T3, skeletal, fast | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7140 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P45378 | |
| TNNT3 | |
| Recombinant protein corresponding to Human TNNT3. Recombinant protein control fragment (Product #RP-104245). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction