missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNNT3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15506182
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15506182 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15506182 Supplier Invitrogen™ Supplier No. PA543681

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: PRPKLTAPKI PEGEKVDFDD IQKKRQNKDL MELQALIDSH FEARKKEEEE Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNNT3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TNNT3
Gene Accession No. P45378
Gene Alias beta TnTF; beta-TnTF; Fast skeletal muscle troponin T; fast skeletal muscle troponin T3; fast skeletal troponin T beta-1 isoform; fetal troponin T 1; fetal troponin T 2; fetal troponin T 3; fTnT; skeletal muscle fast-twitch TnT; skeletal troponin T alpha-1 isoform; Tnnt3; Tnt; TnT1f; TnT2.5f; TnT2f; TnT2fa; TnT4f; tnTf; troponin T 2.5f precursor; troponin T 2f precursor; troponin T 2fa precursor; troponin T 3f; troponin T 4f precursor; troponin T fast skeletal muscle type; troponin T type 3 (skeletal, fast); troponin T, fast skeletal muscle; troponin T1f precursor; troponin T3, fast skeletal type; troponin T3, skeletal, fast; troponin-T3, skeletal, fast
Gene Symbols TNNT3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human TNNT3 (aa 50-99).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7140
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.