missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNNC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560269
This item is not returnable.
View return policy
Description
Immunogen sequence: NADGYIDPEE LAEIFRASGE HVTDEEIESL M Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 97%.
Troponin (Tn), a key protein complex in the regulation of striated muscle contraction, is composed of 3 subunits. The Tn-I subunit inhibits actomyosin ATPase, the Tn-T subunit binds tropomyosin and Tn-C, while the Tn-C subunit binds calcium and overcomes the inhibitory action of the troponin complex on actin filaments. The protein encoded by this gene is the Tn-C subunit.
Specifications
| TNNC2 | |
| Polyclonal | |
| Unconjugated | |
| TNNC2 | |
| skeletal troponin C; STNC; Tncs; Tnnc2; troponin C type 2 (fast); troponin C, fast skeletal; troponin C, skeletal muscle; troponin C2, fast; troponin C2, fast skeletal type | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7125 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P02585 | |
| TNNC2 | |
| Recombinant protein corresponding to Human TNNC2. Recombinant protein control fragment (Product #RP-99339). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction