missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TNN Monoclonal specifically detects TNN in Mouse samples. It is validated for Dot Blot/ELISA/Immunohistochemistry/Immunohistochemistry (Frozen)/Immunoprecipitation/Western Blot.
Specifications
Specifications
| Antigen | TNN |
| Applications | Dot Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | MTn-12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:200, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:200, Dot Blot |
| Formulation | Ascites |
| Gene Alias | tenascin N, tenascin-N, tenascin-W, TN-N, TNW, TN-W |
| Host Species | Rat |
| Immunogen | Partially purified mouse tenascin. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?