missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNN Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98639CL1
This item is not returnable.
View return policy
Description
TNN Polyclonal antibody specifically detects TNN in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| TNN | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 63923 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| tenascin N, tenascin-N, tenascin-W, TN-N, TNW, TN-W | |
| Produced in rabbits immunized with E. coli-derived Human TNN fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction