missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNK1 Monoclonal Antibody (1B5G3;3F11H6)

Description
MA5-15413 targets TNK1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15413 immunogen is purified recombinant fragment of TNK1 (aa451-560) expressed in E. Coli. .
TNK1 tyrosine kinase, non-receptor 1 belongs to the tyrosine protein kinase family and as such is an important regulator of intracellular signal transduction pathways mediating cellular proliferation, survival, and development. TNK1 is highly expressed in fetal tissues and at lower levels in a few adult tissues. TNK1 may function in signaling pathways utilized broadly during fetal development, and more selectively in adult tissues. TNK1 plays a negative regulatory role in the Ras-Raf1-MAPK pathway, and knockout mice have been shown to develop spontaneous tumors, suggesting a role as a tumor suppressor gene.
Specifications
Specifications
| Antigen | TNK1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1B5G3;3F11H6 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | TNK1 |
| Gene Accession No. | Q13470 |
| Gene Alias | CD38 negative kinase 1; Kinase of embryonic stem cells; Kos1; MGC46193; non-receptor tyrosine kinase; non-receptor tyrosine-protein kinase TNK1; TNK1; Tnk1a; Tnk1b; tyrosine kinase non receptor 1; tyrosine kinase, non-receptor 1; tyrosine kinase, non-receptor, 1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?