missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNK1 Monoclonal Antibody (170CT7.5.4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA537485

Product Code. 17107078

  • £454.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TNK1 Monoclonal Antibody for Western Blot

TNK1 tyrosine kinase, non-receptor 1 belongs to the tyrosine protein kinase family and as such is an important regulator of intracellular signal transduction pathways mediating cellular proliferation, survival, and development. TNK1 is highly expressed in fetal tissues and at lower levels in a few adult tissues. TNK1 may function in signaling pathways utilized broadly during fetal development, and more selectively in adult tissues. TNK1 plays a negative regulatory role in the Ras-Raf1-MAPK pathway, and knockout mice have been shown to develop spontaneous tumors, suggesting a role as a tumor suppressor gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TNK1
Monoclonal
0.45 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q99ML2
TNK1
TNK1 recombinant protein.
400 μL
Primary
Mouse
Antibody
IgG1 κ
Western Blot
170CT7.5.4
Unconjugated
TNK1
CD38 negative kinase 1; Kinase of embryonic stem cells; Kos1; MGC46193; non-receptor tyrosine kinase; non-receptor tyrosine-protein kinase TNK1; TNK1; Tnk1a; Tnk1b; tyrosine kinase non receptor 1; tyrosine kinase, non-receptor 1; tyrosine kinase, non-receptor, 1
Mouse
Protein G
RUO
83813
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.