missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNIP3 Polyclonal Antibody

Description
A suggested positive control is human spleen tissue lysate. PA5-20798 can be used with blocking peptide PEP-0912.
The nuclear factor NF-kappa-B plays key roles in development and immunity. ABIN3 (A20-binding inhibitor of NF-kappa-B activation 3), also known as TNFAIP3-interacting protein 3 (TNIP3), is a novel negative feedback regulator of LPS-induced NF-kappa-B activation. ABIN3 is a 39 kDa protein that negatively regulates NF-kappa-B activation in response to TNF and LPS. ABIN3 is highly expressed in brain, thymus, lymph node, lung and fetal liver, with low expression in kidney, bone marrow. Through its interaction with A20, ABIN3 interferes with TRAF2-mediated transactivation signals and NF-kappa-B inhibition is mediated by the ABIN-homology domain 2. ABIN3 has been found to be induced by Listeria infection and can be slightly downregulated by dexamethasone. Enhanced expression of ABIN3 in monocytes is associated with sepsis. Thus, ABIN3 is an IL-10-induced gene product capable of attenuating NF-kappa-B in human macrophages yet is inoperative in mice and represents a basis for species-specific differences in IL-10 actions. At least four isoforms of ABIN3 are known to exist.
Specifications
Specifications
| Antigen | TNIP3 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | TNIP3 |
| Gene Accession No. | Q96KP6 |
| Gene Alias | 9030611K07Rik; A20-binding inhibitor of NF-kappa-B activation 3; ABIN3; ABIN-3; ABIN-3 beta; LIND; Listeria induced; listeria-induced gene protein; TNFAIP3 interacting protein 3; TNFAIP3-interacting protein 3; TNFAIP3-interacting protein 3 beta; Tnip3; TNIP3 beta |
| Gene Symbols | TNIP3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?