missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNIP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103506
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TNIP3.
The nuclear factor NF-kappa-B plays key roles in development and immunity. ABIN3 (A20-binding inhibitor of NF-kappa-B activation 3), also known as TNFAIP3-interacting protein 3 (TNIP3), is a novel negative feedback regulator of LPS-induced NF-kappa-B activation. ABIN3 is a 39 kDa protein that negatively regulates NF-kappa-B activation in response to TNF and LPS. ABIN3 is highly expressed in brain, thymus, lymph node, lung and fetal liver, with low expression in kidney, bone marrow. Through its interaction with A20, ABIN3 interferes with TRAF2-mediated transactivation signals and NF-kappa-B inhibition is mediated by the ABIN-homology domain 2. ABIN3 has been found to be induced by Listeria infection and can be slightly downregulated by dexamethasone. Enhanced expression of ABIN3 in monocytes is associated with sepsis. Thus, ABIN3 is an IL-10-induced gene product capable of attenuating NF-kappa-B in human macrophages yet is inoperative in mice and represents a basis for species-specific differences in IL-10 actions. At least four isoforms of ABIN3 are known to exist.
Specifications
| TNIP3 | |
| Polyclonal | |
| Unconjugated | |
| TNIP3 | |
| 9030611K07Rik; A20-binding inhibitor of NF-kappa-B activation 3; ABIN3; ABIN-3; ABIN-3 beta; LIND; Listeria induced; listeria-induced gene protein; TNFAIP3 interacting protein 3; TNFAIP3-interacting protein 3; TNFAIP3-interacting protein 3 beta; Tnip3; TNIP3 beta | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 414084, 79931 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96KP6 | |
| TNIP3 | |
| A synthesized peptide derived from human TNIP3(Accession Q96KP6), corresponding to amino acid residues T77-L127. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction