missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNIP3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00
Specifications
| Antigen | TNIP3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
TNIP3 Polyclonal specifically detects TNIP3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TNIP3 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| NP_079149 | |
| 79931 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ABIN3. The immunogen is located within the last 50 amino acids of ABIN3. Amino Acid Squence: KQREHPPDYQWYALD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| A20, A20-binding inhibitor of NF-kappa-B activation 3, ABIN3, ABIN-3ABIN-3 beta, FLJ21162, LINDListeria induced, Listeria-induced gene protein, TNFAIP3 interacting protein 3, TNFAIP3-interacting protein 3, TNFAIP3-interacting protein 3 beta, TNIP3 beta | |
| TNIP3 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title