missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112919
This item is not returnable.
View return policy
Description
TNIP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
Specifications
| TNIP1 | |
| Polyclonal | |
| Unconjugated | |
| TNIP1 | |
| A20-binding inhibitor of NF-kappa B activation; A20-binding inhibitor of NF-kappa-B activation 1; ABIN; ABIN1; ABIN-1; AU018810; HIV-1 Nef-interacting protein; KIAA0113; Naf1; Nef; Nef-associated factor 1; Nef-associated factor 1 SNP; nip40-1; TNFAIP3 interacting protein 1; TNFAIP3-interacting protein 1; Tnip1; VAN; Virion-associated nuclear shuttling protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 10318, 363599 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.59 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q15025 | |
| TNIP1 | |
| Recombinant Human TNFAIP3-interacting protein 1 protein (526-636AA). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction