missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNIP1 Polyclonal Antibody

Product Code. 16321835
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16321835 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16321835 Supplier Invitrogen™ Supplier No. PA584883

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GTSSEFEVVT PEEQNSPESS SHANAMALGP LPREDGNLML HLQRLETTLS VCAEEPDHGQ LFTHLGRMAL EFNRLASKVH KNEQRTSILQ TLCEQLRK Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 84%.

Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNIP1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TNIP1
Gene Accession No. Q15025
Gene Alias A20-binding inhibitor of NF-kappa B activation; A20-binding inhibitor of NF-kappa-B activation 1; ABIN; ABIN1; ABIN-1; AU018810; HIV-1 Nef-interacting protein; KIAA0113; Naf1; Nef; Nef-associated factor 1; Nef-associated factor 1 SNP; nip40-1; TNFAIP3 interacting protein 1; TNFAIP3-interacting protein 1; Tnip1; VAN; Virion-associated nuclear shuttling protein
Gene Symbols TNIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TNIP1. Recombinant protein control fragment (Product #RP-107853).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10318
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.