missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFSF13 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10680-1-AP-20UL
This item is not returnable.
View return policy
Description
Members in the TNF superfamily regulate immune responses and induce apoptosis. A novel member in the TNF family was recently identified by several groups and designated APRIL (for A PRoliferation-Inducing Ligand), TALL2 (for TNF and ApoL-related Leukocyte-expressed Ligand 2), and TRDL 1a (for TNF related death ligand 1a) in human and mouse. Two receptors for APRIL were recently identified and designated TACI and BCMA. APRIL stimulates B and T cell proliferation, triggers humoral immune responses, activates NF-kB, and induces cell death. APRIL and its close relative of BlyS and their receptors BCMA and TACI are involved in diseases of autoimmunity and cancer.Specifications
| TNFSF13 | |
| Polyclonal | |
| Unconjugated | |
| TNFSF13 | |
| APRIL, CD256, ligand, TALL 2, TALL2, TNF related death ligand 1, TNFSF13, TRDL 1, UNQ383/PRO715, ZTNF2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8741 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75888 | |
| TNFSF13 | |
| TNFSF13 Fusion Protein Ag1056 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction