missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF8, anti-Human, Clone: Ki-1/1747R, Abnova™
Monoclonal Antibody
Brand: Abnova RAB00370.100ug
This item is not returnable.
View return policy
Specifications
| TNFRSF8 | |
| Monoclonal | |
| Unconjugated | |
| In 10 mM PBS. | |
| CD30/D1S166E/KI-1 | |
| Original antibody is raised against recombinant protein corresponding to full length human TNFRSF8. | |
| RUO | |
| 943 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Ki-1/1747R | |
| Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| tumor necrosis factor receptor superfamily, member 8 | |
| TNFRSF8 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction