missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF6B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24999-1-AP-150UL
This item is not returnable.
View return policy
Description
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.Specifications
| TNFRSF6B | |
| Polyclonal | |
| Unconjugated | |
| TNFRSF6B | |
| DCR3, Decoy receptor 3, Decoy receptor for Fas ligand, M68, TNFRSF6B, TR6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8771 | |
| -20°C | |
| Liquid |
| Immunoprecipitation | |
| 0.47 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O95407 | |
| TNFRSF6B | |
| TNFRSF6B Fusion Protein Ag19781 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction