missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNFRSF14 (HVEM) Chimeric Recombinant Rabbit Monoclonal Antibody (HMHV-1B18)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548130
This item is not returnable.
View return policy
Description
Specificity: This antibody is specific for Herpes Virus Entry Mediator (HVEM, TR2), a type I transmembrane protein of TNF-receptor superfamily.
TNFRSF14 is a member of the TNF-receptor superfamily. TNFRSF14 was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to TNFRSF14 has been shown to be part of the viral entry mechanism. The cytoplasmic region of TNFRSF14 was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. Activation of the signal transduction pathway involving TNFRSF14 in T cells stimulates T cell proliferation and cytokine production, leading to inflammation and enhanced CTL-mediated tumor immunity, suggesting that these proteins may be useful as potential targets for controlling cellular immune responses. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these TNFRSF14 variants have not been determined.
Specifications
| TNFRSF14 (HVEM) Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q80WM9 | |
| TNFRSF14 | |
| This antibody was raised by immunising Armenian hamsters with mouse HVEM:Fc fusion protein. | |
| 200 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG κ |
| Flow Cytometry, Functional Assay, Inhibition Assays, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| HMHV-1B18 | |
| Unconjugated | |
| TNFRSF14 | |
| Atar; CD270; CD40-like protein; herpes virus entry mediator; herpes virus entry mediator A; Herpesvirus entry mediator A; HGNC:11912; HVEA; Hvem; LIGHTR; RP3-395M20.6; sCD2710; TNF receptor superfamily member 14; Tnfrs14; TNFRSF14; TR2; tumo; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2; UNQ329/PRO509 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 230979 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction