Learn More
Invitrogen™ TNFRSF14 Chimeric Recombinant Armenian Hamster Monoclonal Antibody (HMHV-1B18)
Armenian Hamster Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551650
Description
This antibody is specific for Herpes Virus Entry Mediator (HVEM, TR2), a type I transmembrane protein of TNF-receptor superfamily. This receptor, which is expressed on most cell types, including T cells, B cells, monocytes, neutrophils, and dendritic cells. Binding of HSV viral envelope glycoprotein D (gD) to this receptor protein has been shown to be part of the viral entry mechanism. The cytoplasmic region of HVEM was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. HVEM has also been demonstrated to be a unique ligand for BTLA (B and T lymphocyte attenuator). The conservation of the BTLA-HVEM interaction between mouse and human suggests that this system is an important pathway regulating lymphocyte activation and/or homeostasis in the immune response.
TNFRSF14 is a member of the TNF-receptor superfamily. TNFRSF14 was identified as a cellular mediator of herpes simplex virus (HSV) entry. Binding of HSV viral envelope glycoprotein D (gD) to TNFRSF14 has been shown to be part of the viral entry mechanism. The cytoplasmic region of TNFRSF14 was found to bind to several TRAF family members, which may mediate the signal transduction pathways that activate the immune response. Activation of the signal transduction pathway involving TNFRSF14 in T cells stimulates T cell proliferation and cytokine production, leading to inflammation and enhanced CTL-mediated tumor immunity, suggesting that these proteins may be useful as potential targets for controlling cellular immune responses. Multiple alternatively spliced transcript variants have been described, but the full-length nature of some of these TNFRSF14 variants have not been determined.
Specifications
| TNFRSF14 Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q80WM9 | |
| TNFRSF14 | |
| This antibody was raised by immunising Armenian hamsters with mouse HVEM:Fc fusion protein. | |
| 200 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG κ |
| Flow Cytometry, Functional Assay, Inhibition Assays, Immunoprecipitation, Western Blot | |
| HMHV-1B18 | |
| Unconjugated | |
| TNFRSF14 | |
| Atar; CD270; CD40-like protein; herpes virus entry mediator; herpes virus entry mediator A; Herpesvirus entry mediator A; HGNC:11912; HVEA; Hvem; LIGHTR; RP3-395M20.6; sCD2710; TNF receptor superfamily member 14; Tnfrs14; TNFRSF14; TR2; tumo; Tumor necrosis factor receptor superfamily member 14; tumor necrosis factor receptor superfamily, member 14; tumor necrosis factor receptor superfamily, member 14 (herpesvirus entry mediator); Tumor necrosis factor receptor-like 2; tumor necrosis factor receptor-like gene2; UNQ329/PRO509 | |
| Armenian Hamster | |
| Protein A | |
| RUO | |
| 230979 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.