missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNFR2 Recombinant Rabbit Monoclonal Antibody (112), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£36.72 - £93.50
Specifications
| Antigen | TNFR2 |
|---|---|
| Clone | 112 |
| Concentration | 100 μg/mL |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019472
![]() |
Invitrogen™
MA552371 |
100 Tests |
£93.50
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019312
|
Invitrogen™
MA552372 |
25 Tests |
£36.72
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TNFR2 Recombinant Monoclonal antibody specifically detects TNFR2 in Mouse samples. It is validated for Flow CytometrySpecifications
| TNFR2 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.5% BSA and 0.03% ProClin 300 | |
| CD120b; Etanercept; p75; p75 TNF receptor; p75 TNFR; p75TNFR; p80 TNF-alpha receptor; sCD120b; soluble CD120b; soluble TNFR1B variant 1; soluble TNFR2; sTNF R2; sTNFR 2; sTNFR2; TBP-2; TBPII; TNF receptor beta chain; TNF receptor superfamily member 1B; TNF-alphaR2; TNFalpha-R2; TNFBR; TNFR; TNFR II; Tnfr-1; TNFR1B; TNFR2; TNF-R2; Tnfr-2; TNF-R75; TNFR80; TNFRII; TNF-RII; TNF-R-II; TNFR-II; TNFRSF1B; tumor necrosis factor beta receptor; tumor necrosis factor binding protein 2; Tumor necrosis factor receptor; tumor necrosis factor receptor 2; Tumor necrosis factor receptor superfamily member 1B; Tumor necrosis factor receptor superfamily member 1b, membrane form; tumor necrosis factor receptor superfamily, member 1B; tumor necrosis factor receptor type II; Tumor necrosis factor-binding protein 2 | |
| Recombinant Mouse TNFR2/CD120b/TNFRSF1B protein (Met1-Gly258). | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Tnfrsf1b |
| 112 | |
| Flow Cytometry | |
| APC | |
| Rabbit | |
| Mouse | |
| P25119 | |
| 21938 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title