missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TNFR1 (Soluble) Monkey ELISA Kit

Product Code. 15532107
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
96 tests
Pack of 10
This item is not returnable. View return policy

Product Code. 15532107

Brand: Invitrogen™ BMS653TEN

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to monkey sTNF-R I
  • HRP-Conjugate anti-monkey sTNF-R I monoclonal antibody
  • Monkey sTNF-R I Standard concentrate, 12 U/ml upon dilution
  • Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
  • Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
  • Substrate Solution (tetramethyl-benzidine)
  • Stop Solution (1M Phosphoric acid)
  • Blue-Dye
  • Green-Dye
  • Adhesive Films

Sandwich ELISA Kit

The Monkey Tumor Necrosis Factor Receptor 1 (Mk TNFR1) ELISA quantitates Mk TNFR1 in monkey(baboon, cynomolgus) serum or cell culture medium. The assay will exclusively recognize both natural and recombinant Mk TNFR1. Principle of the method The Monkey TNFR1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

Assay Sensitivity 0.06 U/mL
Conjugate HRP
Sample Type Baboon Serum, Cell Culture Supernatant, Cynomolgus Serum
Target Species Monkey
For Use With (Application) ELISA
Quantity 10 x 96 Tests
Synonym CD120a, TNFR60, TNF Receptor superfamily member 1A
Target sCD120a (sTNF Receptor I)

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.