Learn More
TNFR1 Mouse anti-Human, CoraLite 594, Clone: 2A6E3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60192-100UL
Description
The TNFR1 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human TNFR1. This antibody recognizes human, mouse, rat, pig antigen. The TNFR1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| TNFR1 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 7132 | |
| IgG1 |
| Immunofluorescence | |
| 2A6E3 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |