missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFR Related Protein (LTβR) Mouse anti-Human, BV786, Clone: hTNFR-RP-M12, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744682
This item is not returnable.
View return policy
Description
TNFR Related Protein (LTβR) Monoclonal antibody specifically detects TNFR Related Protein (LTβR) in Human samples. It is validated for Flow Cytometry.
Specifications
| TNFR Related Protein (LTβR) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| hTNFR-RP-M12 | |
| Brilliant Violet 786 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction