Learn More
Invitrogen™ TNFAIP8L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589368
Description
Immunogen sequence: MESFSSKSLA LQAEKKLLSK MAGRSVAHLF IDETSSEVLD ELYRVSKEYT HSRPQAQRVI KDLIKVAIKV AVLHRNGSFG PSELALATRF RQKLRQGAMT ALSFGEVDFT FEAAVLAGLL TECRDVLLEL VEHHLTPKSH GRIRHVFDHF SDPGLLTALY GPDFTQHLGK ICDGLRKLLD EGKL; Positive Samples: Mouse thymus, Mouse spleen, Mouse small intestine.
TNFAIP8L2 - Tumor necrosis factor alpha-induced protein 8-like protein 2 (also known as TIPE2) acts as a negative regulator of innate and adaptive immunity by maintaining immune homeostasis. It is a negative regulator of Toll-like receptor and T-cell receptor functions. TIPE2 prevents hyperresponsiveness of the immune system and maintains immune homeostasis. It inhibits JUN/AP1 and NF-kappa-B activation. TIPE2 also promotes Fas-induced apoptosis.
Specifications
| TNFAIP8L2 | |
| Polyclonal | |
| Unconjugated | |
| Tnfaip8l2 | |
| 1810019A08Rik; AW610835; inflammation factor 20; inflammation factor protein 20; RGD1310519; TIPE2; TNF alpha induced protein 8 like 2; TNF alpha-induced protein 8-like protein 2; Tnfaip8l2; TNFAIP8-like protein 2; TP8L2; Tumor necrosis factor alpha-induced protein 8-like protein 2; tumor necrosis factor, alpha induced protein 8 like 2; tumor necrosis factor, alpha-induced protein 8-like 2; tumor necrosis factor, alpha-induced protein 8-like protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 310663, 69769, 79626 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.87 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6AYJ8, Q6P589, Q9D8Y7 | |
| Tnfaip8l2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-184 of human TNFAIP8L2 (NP_0788512). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.