missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFAIP8L2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93009-0.02ml
This item is not returnable.
View return policy
Description
TNFAIP8L2 Polyclonal antibody specifically detects TNFAIP8L2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| TNFAIP8L2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunoprecipitation 1:500 - 1:1000 | |
| FLJ23467, Inflammation factor protein 20, TIPE2, TIPE2inflammation factor 20, TNF alpha-induced protein 8-like protein 2, TNFAIP8-like protein 2, tumor necrosis factor alpha-induced protein 8-like protein 2, tumor necrosis factor, alpha-induced protein 8-like 2, tumor necrosis factor, alpha-induced protein 8-like protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-184 of human TNFAIP8L2 (NP_078851.2). MESFSSKSLALQAEKKLLSKMAGRSVAHLFIDETSSEVLDELYRVSKEYTHSRPQAQRVIKDLIKVAIKVAVLHRNGSFGPSELALATRFRQKLRQGAMTALSFGEVDFTFEAAVLAGLLTECRDVLLELVEHHLTPKSHGRIRHVFDHFSDPGLLTALYGPDFTQHLGKICDGLRKLLDEGKL | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79626 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction