missing translation for 'onlineSavingsMsg'
Learn More

TNFAIP8L1 Antibody, Novus Biologicals™

Product Code. 18330420 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330420 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330420 Supplier Novus Biologicals Supplier No. NBP191434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TNFAIP8L1 Polyclonal specifically detects TNFAIP8L1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TNFAIP8L1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_689575
Gene Alias FLJ39932, MGC17791, TNF alpha-induced protein 8-like protein 1, TNFAIP8-like protein 1, tumor necrosis factor alpha-induced protein 8-like protein 1, tumor necrosis factor, alpha-induced protein 8-like 1, tumor necrosis factor, alpha-induced protein 8-like protein 1
Gene Symbols TNFAIP8L1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human TNFAIP8L1. Peptide sequence AKSHGRINHVFGHLADCDFLAALYGPAEPYRSHLRRICEGLGRMLDEGSL.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126282
Test Specificity Expected identity based on immunogen sequence: Canine: 100%; Chicken: 85%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.