missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNFAIP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585357
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Predicted reactivity: Mouse (83%), Pig (81%), Rhesus Monkey (97%). Positive Control: 3xFlag-human TNFAIP3-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TNFAIP3, also known as A20, is located in chromosome band 6q23, a region that is often deleted in B cell lymphomas. Recently, it was identified as a tumor suppressor gene in Hodgkin lymphoma and several subtypes of non-Hodgkin lymphomas. TNFAIP3 was initially identified as a zinc-finger protein that is rapidly and transiently induced by TNF-a, inhibiting NF-kappa-B-dependent gene expression, and protecting cells from TNF-a-cytotoxicity. Overexpression of TNFAIP3 also inhibits the TLR2- and TLR4-mediated interleukin-8 synthesis in airway epithelial cells, suggesting that TNFAIP3 also acts as a negative regulator of TLR-mediated inflammatory responses, thereby protecting the host against harmful over-responses to pathogens. At least two isoforms of TNFAIP3 are known to exist.
Specifications
| TNFAIP3 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP3 | |
| A20; A20p37; A20p50; AISBL; OTU domain-containing protein 7C; OTUD7C; Putative DNA-binding protein A20; RP11-356I2.3; TNF alpha induced protein 3; TNF alpha-induced protein 3; TNFA1P2; TNFAIP3; TNFAIP3 (A20); Tnfip3; Tumor necrosis factor alpha-induced protein 3; tumor necrosis factor induced protein 3; tumor necrosis factor inducible protein A20; tumor necrosis factor, alpha induced protein 3; tumor necrosis factor, alpha-induced protein 3; zinc finger protein A20 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7128 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.67 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P21580 | |
| TNFAIP3 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human TNFAIP3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction