missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNFAIP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565073
This item is not returnable.
View return policy
Description
Immunogen sequence: RWAEDVPPQR LDGHCHSELA IDIIQITSQA QAKAESITLD LGSQIKRVLL VELPAFLRSY QRAFNEFLER GKQLTNYRAN VIANINNCLS FRMSMEQNWQ VPQDTLSLLL GPLGELKSH.
TNFAIP2, also known as B94, is a TNF-a-regulated gene that is expressed in endothelial cells, peripheral blood cells, and mature sperm. Recently, elevated TNFAIP2 expression was observed in nasopharyngeal carcinoma, and high expression of TNFAIP2 was significantly correlated with higher levels of cell migration, invasion and metastasis, suggesting that TNFAIP2 may serve as a useful prognostic indicator for nasopharyngeal carcinoma. TNFAIP2 has also been implicated as part of the viral-sensing circuit of the innate immune response.
Specifications
| TNFAIP2 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP2 | |
| B94; Exoc3l3; exocyst complex component 3-like 3; M-sec; primary response gene B94 protein; TNF alpha induced protein 2; TNF alpha-induced protein 2; TNF alpha-IP 2; Tnfaip2; tnfb94; Tnfip2; tumor necrosis factor alpha-induced protein 2; tumor necrosis factor induced protein 2; tumor necrosis factor, alpha induced protein 2; tumor necrosis factor, alpha-induced protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7127 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q03169 | |
| TNFAIP2 | |
| Recombinant Human TNFAIP2. Recombinant protein control fragment (Product #RP-105835). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction