missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNFAIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553043
This item is not returnable.
View return policy
Description
Immunogen sequence: FGTILNYLRD DTITLPQNRQ EIKELMAEAK YYLIQGLVNM CQSALQDKKD SYQPVCNIPI ITSLKEEERL IESSTKPVVK LLYNRSNNKY S Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in a tissue-specific manner.
Specifications
| TNFAIP1 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP1 | |
| B12; B61; BACURD2; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2; BTB/POZ domain-containing protein TNFAIP1; BTBD34; EDP1; Edp-1; hBACURD2; Protein B12; TNF alpha induced protein 1; Tnfaip1; Tnfip1; tumor necrosis factor induced protein 1; tumor necrosis factor, alpha induced protein 1; tumor necrosis factor, alpha-induced protein 1 (endothelial); Tumor necrosis factor, alpha-induced protein 1, endothelial; Tumor necrosis factor-induced protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7126 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13829 | |
| TNFAIP1 | |
| Recombinant protein corresponding to Human TNFAIP1. Recombinant protein control fragment (Product #RP-90725). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction