missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TNFAIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101162
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TNAP1.
This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. Studies of a similar gene in mouse suggest that the expression of this gene is developmentally regulated in a tissue-specific manner.
Specifications
| TNFAIP1 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP1 | |
| B12; B61; BACURD2; BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2; BTB/POZ domain-containing protein TNFAIP1; BTBD34; EDP1; Edp-1; hBACURD2; Protein B12; TNF alpha induced protein 1; Tnfaip1; Tnfip1; tumor necrosis factor induced protein 1; tumor necrosis factor, alpha induced protein 1; tumor necrosis factor, alpha-induced protein 1 (endothelial); Tumor necrosis factor, alpha-induced protein 1, endothelial; Tumor necrosis factor-induced protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21927, 287543, 7126 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70479, Q13829, Q7TNY1 | |
| TNFAIP1 | |
| A synthesized peptide derived from human TNFAIP1(Accession Q13829), corresponding to amino acid residues E103-S153. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction